Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Hazardous Material Pressure Sensitive Numbers for Labeling
Specifications
Color | Black, White |
---|---|
Format | Label |
Material | Vinyl |
Product Type | Label Number |
For Use With (Application) | Hazardous materials |
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
17-981-58A
|
National Marker™
DCN 4 0 |
Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||||
17-981-58B
|
National Marker™
DCN 4 1 |
1 | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
17-981-58C
|
National Marker™
DCN 4 2 |
2 | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
17-981-58D
|
National Marker™
DCN 4 3 |
3 | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
17-981-58E
|
National Marker™
DCN 4 4 |
4 | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Specifications
Black, White | |
Vinyl | |
Hazardous materials | |
0.0045 mil | |
5 in. |
Label | |
Label Number | |
5/Pk. | |
5 in. | |
12.7 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title