Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Danger: Follow Confined Space Entry Procedure Before Signs
$21.20 - $37.70
Specifications
Color | Black/Red on White |
---|---|
Message | DANGER Follow Confined Space Entry Procedure Before |
Certifications/Compliance | OSHA 29 CFR 1910.145 |
Quantity | 1 Ea. |
For Use With (Application) | Perfect way to alert employees and guests when a hazard is present in your facility |
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Price | Quantity & Availability | ||||
18-999-731
|
National Marker™
D277RB |
25.4 x 35.6 cm | Plastic |
Each for $37.70
|
|
18-999-730
|
National Marker™
D277R |
7 x 10 in. | Plastic |
Each for $28.50
|
|
18-999-732
|
National Marker™
D277P |
7 x 10 in. | Vinyl |
Each for $21.20
|
|
Specifications
Black/Red on White | |
OSHA 29 CFR 1910.145 | |
Perfect way to alert employees and guests when a hazard is present in your facility |
DANGER Follow Confined Space Entry Procedure Before | |
1 Ea. | |
OSHA sign |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title