Learn More
Yamato Scientific America, Inc. Yamato 74L Natural Convection Oven, 115V
Supplier: Yamato Scientific America, Inc. DX402C
Standard 74L natural convection constant temperature drying oven with extensive features and simple operation. Economical and cost saving. Easy to use and maintain. Excellent temperature accuracy. Digital PID controller, calibration off-set and increased safety and self diagnostic functions. Operating temp ambient +5~300°C