Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Naspm trihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Selective antagonist of Ca2+-permeable AMPA receptors; blocks AMPA receptors lacking the GluA2 subunit. Protects hippocampal neurons against global ischemia-induced cell death.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 479.91 |
Percent Purity | >97% |
Chemical Name or Material | Naspm trihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction