Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Nanotrap Enhancement Reagent 2
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Use of Nanotrap Enhancement Reagents with Nanotrap Microbiome Particles improves the binding of microbes in wastewater samples, further enhancing downstream detection of nucleic acids.
$108.00 - $260.00
Specifications
Isolation Technology | Magnetic Particles |
---|---|
Sample Type | Wastewater |
Sequencing Type | RT-PCR and Digital PCR Sequencing |
Product Type | Enhancement Reagent 2 |
Form | Liquid |
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
23-285-974
![]() |
Streck
250071 |
10 mL |
Each for $108.00
|
|
|||||
23-285-975
![]() |
Streck
250073 |
30 mL |
Each for $260.00
|
|
|||||
Description
- Utilize affinity capture and enrichment to attract and bind low abundance viruses.
- Can be used with clinical swab samples, saliva, urine, wastewater, food wash and blackwater.
- Compatible with PCR, sequencing or immunoassay analysis methods.
- Designed to capture and concentrate a variety of microbes.
- Increase sensitivity of existing SARS-CoV-2 assay by capturing and concentrating whole virus.
- Nanotrap Enhancement Reagents improve microbe detection in wastewater samples by at least 1-2 Ct values.
Specifications
Magnetic Particles | |
RT-PCR and Digital PCR Sequencing | |
Liquid |
Wastewater | |
Enhancement Reagent 2 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title