Learn More
Cytiva Nanosep MF Centrifugal Devices with Bio-Inert Membrane - 0.45 µm, wildberry (24/pkg)
Supplier: Cytiva ODM45C33
Nucleic acid binding (NAB) Nanoseps have a unique dual layer glass fiber membrane allowing isolation of all nucleic acids This product is not approved for use in medical, clinical, surgical or other patient applications.