Learn More
Izon Science Usa Ltd Nanopore (50-330nm)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Izon Science Usa Ltd NP100
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This item has a minimum order quantity of 3 per supplier requirements
Nanopores (NP) are conical holes formed in flexible polyurethane membranes. On the qNano and qViro-X platforms, these can be stretched to optimise the nanopore size to the particles being measured.