Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH, Alexa Fluor 488 [CTK0101, CTK0102]
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Proteintech Group Inc SRBAF4881100
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Nano-Secondary alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
This item is not returnable.
View return policy