Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile

Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.

Supplier:  Thermo Scientific™ 7282045

 View more versions of this product

Catalog No. 09-740-120

  • $2,380.65 / Case of 500

    Save 22%

    Reg : $3,060.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

Description

Description

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTFE
Non-sterile
Fewer resources used and less waste
Nalgene™
10 to 100 mL (Sample)
25 mm
0.45 μm
Filtration of HPLC/GC samples and solvents, sterile venting
Polypropylene (Housing)
Syringe Filter
25 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit