Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Filter Storage Bottles
Save time and minimize contamination with these radiation-sterilized filter storage bottles. Excellent as single-use storage containers.
Supplier:  Thermo Scientific™ 4550250
 View more versions of this product
Catalog No. 09-740-25E
  • $140.65 / Case of 24
    Save 23%
    Reg : $182.30
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Description
Description
Save time and minimize contamination with gamma-sterilized Thermo Scientific™ Nalgene™ Rapid-Flow™ Filter Storage Bottles. Use with Nalgene Bottle-Top filters or as replacement/extra receptacles for filter units. Excellent as single-use storage containers.
  • Headspace allows for extra serum or other additives to be added to the filtrate
  • Leakproof† screw caps to eliminate pH shift in bottle for longer life of stored media
  • Sterilized by gamma radiation and individually bagged for a 5-year sterile shelf life
  • Non-pyrogenic and non-cytotoxic
  • Available with Fit for Purpose Sterile documentation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Virgon Clear Polystyrene Bottle, White Pigmented Virgin HDPE Closure
Wide
24
250 mL
Sterile
Wide
24
Sterile
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.