Learn More
Thermo Scientific™ Nalgene™ Polypropylene Vacuum Flask PROMO
Replace your glass flask with this Polypropylene Vacuum Flask, which has been molded in one piece for full vacuum service with angled tubulation to prevent tipping.
Supplier: Thermo Scientific™ 1018251S24
Description
- Tubulation at 45° angle prevents tipping when hose is attached
- Barbed adapter accepts 1/4 to 3/8 in. I.D. tubing (tubing sold separately)
- Oval marking area for ease of labeling
- Neck sized to fit standard stopper sizes (stopper not included)
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific™ encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
57.4 oz. | |
Polypropylene | |
Autoclavable |
1.7 L | |
1 Flask/Pk. | |
Stopper No. 10 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.