Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Combinethese flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Supplier: Thermo Scientific™ 41020500
Description
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
- Translucent wall makes liquid level easily visible
- 2000mL size has molded-in graduations
- Mouth sized to accept standard sized stoppers (stopper sold separately)
- Repeatedly autoclavable for long life and rigorous use
Specifications
16 oz. | |
500 mL | |
12 | |
Autoclavable | |
No | |
Non-sterile | |
Stopper No. 7 |
Flask Erlenmeyer PP 500mL | |
PPCO | |
4 | |
Reusable | |
Narrow | |
Plain |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction