Learn More
Sklar Instruments Surgi-OR Nail Nipper, Sklar Angled, Concave Blade, 5-1/2 in., Stainless Steel. EA.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Sklar Instruments 95484
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Surgi-OR Nail Nipper, Sklar. Angled, Concave Blade, 5-1/2 in., Stainless Steel. The Surgi-OR Nail Nipper is used for removing portions of the toe nail or the nail plate. This product features double-spring, partially cross-serrated, plier handles with a locking mechanism that is released upon squeezing the handles. This lock is intended to keep the sharp blades closed when the instrument is not being used. This product is 5-1/2 inches long and features angled, concave blades.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.