Learn More
Sklar Instruments Nail Nipper, Sklar 4-1/2 in., Concave, Smooth, Pliers with Barrel Spring / Ratchet, Stainless Steel. EA.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Sklar Instruments 98485
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Nail Nipper, Sklar. 4-1/2 in., Concave, Smooth, Pliers with Barrel Spring / Ratchet, Stainless Steel. The Merit Nail Nipper is used for removing portions of the toe nail or the nail plate. This product features barrell spring, grooved, plier handles with a locking mechanism that is released upon squeezing the handles. This lock is intended to keep the sharp blades closed when the instrument is not being used. The barrel spring allows for the jaws to open wider for a more accurate placement of the sharp blades. This product is 4-1/2 inches long and features concave blades.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.