Learn More
Perkin Elmer US LLC Nafion Drying Tube for SMS100 Instrument
Supplier: Perkin Elmer US LLC N9309021
This Nafion Drying Tube is included in the Basic Spares Kit (N9309109) and the Extended Parts Spares Kit (N9309110). The SMS 100 is a dedicated mercury analyzer for the determination of total mercury in solid and liquid samples using the principle of thermal decomposition, amalgamation and atomic absorption. The SMS 100 uses a decomposition furnace to release mercury vapor instead of the chemical reduction step used in traditional liquid-based analyzers. Both solid and liquid matrices can be loaded onto the instruments autosampler and analyzed without acid digestion or sample preparation prior to analysis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.