Learn More
Research Products International Corp NAD [B-NAD] [B-Nicotinamide Adenine Dinucleotide, Oxidized free acid hydrate], 1 Gram
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp N301101.0
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
B-Nicotinamide-Adenine Dinucleotide, Oxidized Form (B-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.