Learn More
Hanna Pharmaceutical Supply Co., Inc. 0.9% SODIUM CHLOR.IRR 500ML
Supplier: Hanna Pharmaceutical Supply Co., Inc. R520101
PIC™ is a PVC-free, DEHP-free Plastic Irrigation Container Bottle is designed for easy non-drip pouring Bottle has a molded hand grip for easier container handling The closure is sterile with recessed set port Threaded screw cap covers for the sterile non-drip pouring lip It is recommended that the product be stored at room temperature (25C); however, brief exposure up to 40C does not adversely affect the product 16/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.