Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Dimethylformamide, Biotech Grade, ≥99.9%, Honeywell Burdick & Jackson™
Click to view available options
Quantity:
1 L
2 L
6 x 1 L
Packaging:
Glass Bottle
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 7 (at 20°C) |
Melting Point | -61°C |
Color | Colorless |
Density | 0.944g/mL |
Boiling Point | 152°C to 153°C |
Quantity | 6 x 1 L |
Explosion Limit | 0.152 |
Flash Point | 58°C (136.4°F) |
Ignition Point | 833°F |
Odor | Weak |
Show More |
Safety and Handling
Hazard Statement | P201-P280-P305 + P351 + P338-P308 + P313 |
DOTInformation | 3/PGIII |
RTECSNumber | 200-679-5 |
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction