Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide (Reagent), Fisher Chemical

Catalog Number O24344
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
pH 4
Melting Point -20°C
Color Colorless
Density 0.937g/cm³
Boiling Point 164°C
Quantity 4 L
Flash Point 70°C (158°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3CON(CH3)2
Vapor Pressure 1.7mbar at 25°C
Viscosity 1.02 mPa/s at 20°C
UN Number 2810
Formula Weight 87.12
Grade Reagent
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0
DOTInformation Class 6.1: Poison
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit