Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-1-Hexane sulfonic acid, Sodium Salt, Monhydrate, 99.3 %, MP Biomedicals™
Click to view available options
Quantity:
10 g
Specifications
Quantity | 10 g |
Appearance | White |
Formula Weight | 188.2 |
Physical Form | Crystalline Powder |
Chemical Name or Material | n-1-Hexanesulfonic Acid Sodium Salt Monhydrate |