Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Gas Phase Filters
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful fumes and odors.
Supplier: Mystaire™ MY2807
Description
- Manufactured with high-grade coconut shell carbon
- Activated carbon filter that provides more surface area for a higher degree of absorbency
- Chemically impregnated carbon for increased load capacity
Specifications
Carbon | |
Aura Elite 42 Ductless Fume Hood | |
Gas Phase Filter | |
Formaldehyde |