Learn More
Thomas Scientific MyFuge 12 Mini Centrifuge, 100-240V (US Plug)
Supplier: Thomas Scientific 1220P38
Unlike traditional mini centrifuges, the MyFuge 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor is all that is required for running 12 microtubes and 4 PCR strips simultaneously.