Learn More
MP Biomedicals™ Mycoplasma Hoechst Stain Kit
The Mycoplasma Hoechst Stain Kit is designed for rapid in situ detection of mycoplasma, and other prokaryotic organisms, in cell cultures in less than two hours.
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN93030001
|
MP Biomedicals™
093030001 |
20 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
ICN3030000
|
MP Biomedicals™
093030000 |
100 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- This rapid mycoplasma detection procedure is based on the Hoechst Stain method cited by the Tissue Culture Association (TCA Procedure No, 75361.).
- Several DNA fluorochromes such as DAPI, quinacrine mustard and quinacrine dihydrochloride have been used in the same technique but none of the fluorochromes perform as well as the Hoechst stain with respect to fluorescent effect, slow quenching and minimum background fluorescence.
- The Hoechst Stain method selectively binds minor grooves of the DNA, which allows for efficient staining of the nuclei.
Specifications
Mycoplasma Detection |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.