Learn More
Lonza Walkersville MycoAlert Mycoplasma Detection Kit (100 Tests)
Supplier: Lonza Walkersville LT07318
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells. Benefits:Convenient & fastSimply add two reagents to the culture supernatantPerform two luminescence readingsResults in less than 20 minEasy interpretation of results Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma except Ureaplasma)Negligible interference with media componentsContent:1 x 10 mL MycoAlert Reagent (lyophilized)1 x 10 mL MycoAlert Substrate (lyophilized)1 x 20 mL MycoAlert Assay Buffer
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.