Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mystaire™ MY Model PCR Prep Station Class 100 Enclosure
SDP

Compact laminar flow enclosures from Mystaire™ reduce the risk of sample contamination while performing PCR.

Supplier:  Mystaire™ MYPCR48BLITZ

 View more versions of this product

Catalog No. 15-338-960PM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

Built-in 254 nanometer shortwave lamp effectively decontaminates the work area between amplifications. Included safety interlocks prevent operator exposure to potentially harmful UV radiation.

  • Offers class 100 vertical laminar flow air with 15-minute UV light timer
  • Constructed of clear, polycarbonate with chemically impervious polypropylene work surface
  • Features a shelf stand with built-in four-position pipet holder
  • Optional electrical cord access port allows 110V or 220V AC
  • Nominal depth of 24 in. (61 cm)

Specifications

Specifications

PCR Prep Station
Polycarbonate
24 x 48 x 30 in.
762 mm
24 in.
Fluorescent, UV
23 x 47 x 18 in. (584 x 1194 x 457 mm)
30 in.
254 mm
610 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit