Learn More
Mystaire™ MY Model PCR Prep Station Class 100 Enclosure Pre-filter
For use with MY Model PCR prep station class 100 enclosures. Effectively removes submicron particulates.
Supplier: Mystaire™ MYPRELF
Description
- Electrostatically charged fiber filter
- Designed to protect from submicron particulates
Specifications
Particulate | |
1 in. | |
20 in. | |
457 x 508 x 25 mm (18 x 20 x 1 in.) | |
457 mm | |
Prefilter |
32 in. wide MY Model PCR Prep Station Class 100 Enclosure | |
18 in. | |
25 mm | |
12 Pk. | |
508 mm | |
Electrostatically Charged Fiber |