Learn More
Teledyne ISCO, Inc Mx Class Single Piston Pump With Eccentric Drive And Shaped Cam; 0.00 - 10.00 Ml/Min. ; 5,000 Psi
Supplier:  Teledyne ISCO, Inc MX010SRX
Encompass
MX Class Single Piston Pump with Eccentric Drive and Shaped Cam; 0.00 - 10.00 mL/min. ; 5,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9955