Learn More
Eisco™ Muscovite Mica Mineral Specimen
Muscovite Mica Mineral Specimen forms in thin sheets that are individually weak, but comprehensively strong.
$3.00 - $30.75
Catalog Number | Mfr. No. | Specimen Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Specimen Size | Quantity | Price | Quantity & Availability | ||||
S24704
|
Eisco™
ESNG0022 |
2 to 3 cm | 1 Ea. |
Each for $3.00
|
|
||||
S26598
|
Eisco™
ESNG0022PK10 |
2 to 3 cm | 10/Pk. |
Each for $6.45
|
|
||||
S24705
|
Eisco™
ESNG0022PK12 |
2 to 3 cm | 12/Pk. |
Each for $30.75
|
|
||||
S26657
|
Eisco™
ESNG0022HAND |
7 to 8 cm | 1 Ea. |
Each for $21.75
|
|
||||
Description
- Unique thin layered structure
- For use in classroom identification labs and exercises
- For use in study of variable hardness and cleavage
- Correlates well to Next Generation Science Standards Evidence Statements
- Pair with biotite mica to demonstrate effect of slight variations in mineral's chemistry
Specifications
Mineral, Muscovite Mica |