Learn More
Electron Microscopy Sciences QuantomiX WetSEM MultiWell Aspirator
Supplier: Electron Microscopy Sciences MA4
Multi-well Aspirator system is designed to safely and conveniently aspirate liquids from the QX-102 capsules, and is required for applications that need liquid exchange in capsules during sample preparation or cell culturing.