Learn More
HANNA INSTRUMENTS INC MULTIPARAMETER PHOTOMETER KIT
Supplier: HANNA INSTRUMENTS INC HI97115UC
The HI97115UC Kit is a compact and versatile Marine multiparameter photometer designed to accurately determine pH, Alkalinity, Ammonia, Calcium, Magnesium, Nitrate, Nitrite, and Phosphate levels in aquariums and marine biology applications. The photometer is suitable for field and bench measurements.The HI97115 can be used as a stand-alone photometer or can be connected to the Hanna Lab App with a compatible smart device via the integrated Bluetooth® module. When connected, Hanna Lab App functions include measurement with the ability to add notes, data logging with extended storage capacity, data sharing, and the ability to create and save method groups.No warm up time needed before taking a measurement.Tutorial mode for easy step-by-step instructions.CAL CheckTM for performance verification and calibration
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.