Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKA™ MultiDrive BL 2000 Blender Vessel
![SDP](/content/dam/fishersci/glyphs/sdp.png)
BL 2000 is a vessel with mixing knife and is particularly useful in combination with IKA MultiDrive basic for wet grinding of a variety of applications, such as meat, sausage, soy beans, silage, vegetables.
Supplier: IKA™ 25003961
This item is not returnable.
View return policy
Description
- All parts can be cleaned in a dishwasher. The knife is easy to dismantle for intensive cleaning.
- The scope of delivery includes a standard mixing knife made of stainless steel.
Specifications
Blender Vessel | |
MultiDrive | |
Stainless Steel |
2000 mL | |
5 Mohs | |
Max: 20 mm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction