Learn More
Honeywell™ Multi-purpose Defender Cartridge with HEPA Filter
The Multipurpose Defender Cartridge with HEPA filter is designed to be used with the PA700 PAPR.
Supplier: Honeywell™ PA7DEHE
Description
- PA700 PAPR Multipurpose Cartridge with HEPA filter: [Organic Vapor, Acid Gas (Chlorine, Hydrogen Chloride, Sulfur Dioxide, Hydrogen Fluoride, Chlorine Dioxide, Hydrogen Sulfide)], Ammonia, Methylamine, Formaldehyde, with High Efficiency Particulate Air-Purifying (HEPA) filter
Specifications
COMBINATION | |
Olive Green | |
Honeywell PA700 Powered Air Purifying Respirator (PAPR) | |
4/Box |
Multigas | |
PAPR Filter Cartridge | |
PA700 PAPR Series Filter Cartridge |