Learn More
Electron Microscopy Sciences Multi Pin Stub Adapter, JEOL
Supplier: Electron Microscopy Sciences 7595302J
This multi pin stub adapter for JEOL is designed to save time. It is made from vacuum aluminum with stainless steel set screws. It accommodates up to four standard 12.7 mm (1/2") pin stubs with 3.2 mm (1/8") pin.
Type: JEOL