Learn More
Electron Microscopy Sciences Multi Holder Multi Holder for 6 Pin Stubs, Hitachi M4
Supplier: Electron Microscopy Sciences 7595325H
This 42 mm in diameter multi pin holder is designed to save time It is made from machined aluminum with stainless steel set screws. It accommodates up to six standard 12.7 mm (1/2") pin stubs with 3.2 mm (1/8") pin.
Type: Hitachi M4