Learn More
Liofilchem™ MTS™ Ceftobiprole [BPR] 0.002-32 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
Specifications
Antimicrobial Agents | Ceftobiprole |
---|---|
Concentration | 0.002-32 μg/mL |
Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
Description | Liofilchem™ MTS™ Ceftobiprole |
Antibiotic Code | BPR |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
22-777-944
|
Liofilchem™
921401 |
10/Pk. | Individually Packed |
Pack of 10 for $49.99
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-777-942
|
Liofilchem™
92140 |
30/Pk. | Individually Packed |
Pack of 30 for $130.93
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-777-943
|
Liofilchem™
921400 |
100/Pk. | Canister Pack |
Pack of 100 for $411.08
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Specifications
Ceftobiprole | |
-20°C or 2°C to 8°C until the given expiry date | |
BPR | |
Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
0.002-32 μg/mL | |
Liofilchem™ MTS™ Ceftobiprole | |
MIC Test Strip |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.