Learn More
Adam Equipment MTB20 Baby Scale 44lb / 20kg capacity x 0.005lb / 0.005kg readability
Supplier:  Adam Equipment MTB 20
Encompass_Preferred
Versatile and portable, the Adam Equipment MTB baby scale offers excellent value for pediatricians and other healthcare providers. With a capacity of 44lb/20kg, this scale boasts the ability to weigh both newborn infants and toddlers on the same instrument. The MTB features quick start-up and easy operation, so minimal training is needed. A cleverly designed ergonomic cradle provides comfortable support for babies and locks securely in place. The cradle removes easily to reveal a non-slip rubber mat underneath, simplifying the task of weighing toddlers. Suitable for maternity wards, doctors offices and pediatricians. High-contrast display is easily visible from a distance. Simple operation allows quick startup and use with little or no training. Lightweight design simplifies transport and portability.Operates with AC adapter (included) or batteries (not included)
Catalog No. NC2611841