Learn More
Scale Systems MT PFA261 FLOOR SCALE COMBO
Supplier: Scale Systems 30114510
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Mettler Toledo PFA261 Floor Scale Combo 2,500lb x .5lb Capacity, 48”x 48” Platform, Carbon Steel Tread Plate ConstructionThreaded Ball-In-Cup Foot, IND236 Scale Terminal Included