Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Variable-Speed Drive with Remote I/O; 300 rpm
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0752820
Easy-to-use drives offer precise speed control and digital display of rpm¿ideal for metering applicationsFlow rate 0.06 to 3400 mL/min (depends on drive rpm and tubing size)Three digit LED display shows motor rpm ¿ confirm speed setting at a glanceReversible motor¿purge tubing before or after pumping; pump fluid in either directionLED indicators confirm motor directionPowerful motors let you stack 2 or 4 pump heads foINCLUDES6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.