Learn More
MSA™ Toxgard II Gas Monitor: Sensor Kit, O2
This is a kit containing the MSA 10019727 oxygen sensor. These sensors are typically used in oxygen deficiency air monitoring, are normally exposed to ambient conditions, and are not operated below 2% oxygen concentration for extended time periods.
Supplier: MSA™ 813723/DISCBYPRODMGMT
Description
These sensors also compensate for changes in barometric pressure and can be used to monitor for oxygen purity.Specifications
Oxygen | |
Sensor Kit, Oxygen | |
0 to 25% |