Learn More
MSA™ Quick-Disconnect Adapter Accessory, Ring
Accessory used to connect air supply hoses to the manifold and air source. Locking-type quick disconnects are available to interconnect lengths of air supply hose.
Supplier: MSA™ 637830
Description
- Accessory for use with supplied-air respirators
Specifications
637830 | |
Ring |