Learn More
MSA™ PortAire™ Portable Air-Supply Systems
For use with pressure-demand supplied-air respirators
Supplier: MSA™ 807052
Safety Services Available
Overview
Repair/Recertification
Description
- Holds two standard SCBA air cylinders
- Includes compact air-pressure regulating system, anodized aluminum frame and hose
Specifications
Pressure Demand Supply Air Respirator | |
Compact air-pressure regulating system, anodized aluminum frame, hose | |
Portable Air Supply Unit |
2 standard SCBA air cylinders | |
807052 |