Learn More
MSA™ Advantage™ 420 Half-Mask Respirator
One-piece UniBond over-molded silicone facepiece helps eliminate possible leak paths
Supplier:  MSA™ 10119576
 View more versions of this product
Catalog No. 17-200-086
Add to Cart
Description
Description
  • Drop-down harness position allows respirator to be removed while retaining head protection
  • One-piece ComforTop™ cradle headband eliminates pressure points
  • AnthroCurve™ multi-ethnic face seal improves fit-testing among diverse workforce
  • Lock-down harness position allows a personal fit without further user adjustments
  • No loose or dangling straps to impede operation or vision
  • Latex-free; high-quality silicone provides greater durability and product life
  • Textured inner facepiece seal minimizes slippage, improves fit
  • Bayonet and threaded cartridge options allow use of all MSA cartridges
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
Meets OSHA 29 CFR Part 1920.134
Specifications
Specifications
NIOSH 42CFR84
420 Half Mask Respirator Assembly
Asbestos abatement, painting, sanding, grinding, welding
Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve, 2 threaded cartridge adapters
Advantage 420 Respirator Threaded Cartridge Assembly
Silicone
Small
Respiratory Systems
Yes
Product Suggestions
Product Suggestions