Learn More
Malvern Panalytical Ms3000 Dry Windows
Supplier: Malvern Panalytical MAZ2051
Pair of anti-reflection coated windows and Viton window seals for the Mastersizer 3000 dry measurement cell (MAZ2050).