Learn More
Brady™ MRO Plus™ Absorbent Rolls
Strong, durable and low linting, making them the perfect go-to products for nearly every industrial setting
Supplier: Brady™ MRO12P
Description
- Highly versatile, gray absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
- Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
- Medium weight - optimal for most general maintenance needs
- 3-ply Meltblown Polypropylene construction is designed for general industrial applications
- Non-perforated absorbents offer the greatest absorbency and better tear resistance in high traffic areas
- Perforations provide cost control and reduce waste by allowing you to use only what you need
Specifications
12.5 gl/gs | |
Perforated every 12 in., Dimpled, Medium-Linting | |
Polypropylene | |
1 Ea. | |
12 in. x 150 ft. (30.5 cm x 45.2 m) |
Gray | |
20 gal. | |
Overspray leaks and drips, workstations, heavy clean-up and wipe-up, liquid storage drums, outfalls and discharge ponds | |
Heavyweight |