Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ MRO Plus™ Absorbent Rolls

Strong, durable and low linting, making them the perfect go-to products for nearly every industrial setting

Supplier:  Brady™ MRO315PBAG

 View more versions of this product

Catalog No. 17-111-011

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Highly versatile, gray absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • Medium weight - optimal for most general maintenance needs
  • 3-ply Meltblown Polypropylene construction is designed for general industrial applications
  • Non-perforated absorbents offer the greatest absorbency and better tear resistance in high traffic areas
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
Specifications

Specifications

Gray
20 gal.
Polypropylene
1 Roll
150 ft. x 15 in.
38.1 cm
Perforated every 18 in., Dimpled, Medium-linting
75.7 L
MRO Plus Universal All-Purpose Heavy Absorbent Roll
Medium-weight
45.72 m
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit