Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Protamine Sulfate
From salmon sperm
$61.50 - $1091.00
Specifications
Content And Storage | Room Temperature |
---|---|
Format | Powder |
For Use With (Application) | Antiheparin activity, refining of vaccines, removal of DNA from Protein samples |
Product Type | Protamine Sulfate from Salmon Sperm |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN19472901
|
MP Biomedicals™
0219472901 |
1000 mg |
Each for $61.50
|
|
|||||
ICN19472905
|
MP Biomedicals™
0219472905 |
5000 mg |
Each for $118.30
|
|
|||||
ICN19472910
|
MP Biomedicals™
0219472910 |
10,000 mg |
Each for $190.90
|
|
|||||
ICN19472925
|
MP Biomedicals™
0219472925 |
25000 mg |
Each for $347.50
|
|
|||||
ICN19472980
|
MP Biomedicals™
0219472980 |
100,000 mg |
Each for $1,091.00
|
|
|||||
Description
Cell Culture Reagent. Highly purified. Nitrogen: Approx. 22.5%. Hygroscopic white powder.
Specifications
Room Temperature | |
Powder | |
Antiheparin activity, refining of vaccines, removal of DNA from Protein samples | |
Protamine Sulfate from Salmon Sperm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title