Learn More
Cole-Parmer MP-250D-F Standard Digital Melting Point Apparatus, ±0.1°C Temperature Resolution; 100-240 VAC
Supplier:  Cole-Parmer 0301051
Encompass_Preferred
The MP-250D series digital melting point apparatus allows you to easily view three capillary tube samples simultaneously through the 8x magnifying lens. The adjustable extension arm can be set to different viewing heights making it ideal for multiple users. A rotating viewing head and safety eye piece eliminate glare on the lens and protect you against hot zone. A 13-key membrane keypad offers easy-to-use menus. The conveniently located push button controls allow temperature recording without looking away from the sample. A digital microprocessor with 0.1C resolution provides fast warm-up and accurate temperature control. Units can be calibrated by on site or by the manufacturer/service organization. A calibration kit is required if you choose to calibrate on site.
Catalog No. NC2226730