Learn More
Clontech Labs 3P MOUSE UNIVERSAL RNA
Supplier: Clontech Labs 3P 636657
Normalization control for gene expression arrays and quantitative PCR studies. A mixture of several tissues provides high representation compared to universal reference made from cell lines.