Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse/Rat Leptin Quantikine ELISA Kit
solid phase ELISA designed to measure mouse or rat Leptin in cell culture supernates, tissue homogenates, serum, and plasma
$736.60 - $4199.20
Specifications
Assay Range | 31.30 to 2000 pg/mL |
---|---|
Assay Sensitivity | 5.56 pg/mL |
Sample Type | Cell Culture Supernatant, Mouse Plasma (EDTA), Mouse Plasma (Heparin), Mouse Serum, Rat Plasma (EDTA), Rat Plasma (Heparin), Rat Serum, Tissue Homogenates |
Specificity | Natural and Recombinant Mouse and Rat Leptin |
Target Species | Mouse, Rat |
Specifications
31.30 to 2000 pg/mL | |
Cell Culture Supernatant, Mouse Plasma (EDTA), Mouse Plasma (Heparin), Mouse Serum, Rat Plasma (EDTA), Rat Plasma (Heparin), Rat Serum, Tissue Homogenates | |
Mouse, Rat | |
3952 | |
FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS |
5.56 pg/mL | |
Natural and Recombinant Mouse and Rat Leptin | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
Leptin/OB |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title