Learn More
Zymo Research Corporation Mouse Liver DNA (5 ug)
SDP
Supplier:  Zymo Research Corporation D50193
Encompass_Preferred
This genomic DNA set is an ideal control for detection and/or quantification methods against 5-methylcytosine ( 5mC) and 5- hydroxymethylcytosine ( 5hmC) as both modified cytosines are present at physiologically relevant levels and loci. The levels of 5- methylcytosine and 5-hydroxymethylcytosine within the Mouse 5hmC & 5mC DNA Set have been precisely quantified by mass spectrometry (LC/MS) and been validated to be consistent by ELISA.
Catalog No. 50-125-1535